Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST6GALNAC2 Recombinant Rabbit Monoclonal Antibody (025)
GREENER_CHOICE

Catalog No. PIMA531252
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531252 Supplier Invitrogen™ Supplier No. MA531252
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse ST6GALNAC2.

ST6GALNAC2 belongs to a family of sialyltransferases that add sialic acids to the nonreducing ends of glycoconjugates. At the cell surface, these modifications have roles in cell-cell and cell-substrate interactions, bacterial adhesion, and protein targeting (Samyn-Petit et al., 2000 [PubMed 10742600]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST6GALNAC2
Applications ELISA
Classification Recombinant Monoclonal
Clone 025
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ST6GALNAC2
Gene Accession No. P70277
Gene Alias (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase; (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase II; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase II (); Gal-beta-1,3-GalNAc alpha-2,6-sialyltransferase; GalNAc alpha-2,6-sialyltransferase II; II; SAITL1; sialyltransferase 7; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-beta-galactosyl-1,3)-N-acetyl galactosaminde alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) B; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B; sialyltransferase 7B; sialyltransferase-like 1; Siat7; SIAT7B; SIAT7-B; SIATL1; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6 GalNAc alpha-2,6-sialyltransferase 2; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6GalNAc; ST6GalNAc II; St6galnac2; ST6GALNAC2, alpha-2,6-sialyltransferase 2; ST6GalNAcII; ST6GalNAII; SThM
Gene Symbols ST6GALNAC2
Host Species Rabbit
Immunogen Recombinant Mouse ST6GALNAC2 protein (Ser28-Arg373).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20446
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.