Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ST6GALNAC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543295
Description
Peptide sequence: QLTRMYPGLQ VYTFTERMMA YCDQIFQDET GKNRRQSGSF LSTGWFTMIL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein prefers glycoproteins rather than glycolipids as substrates and shows restricted substrate specificity, utilizing only the trisaccharide sequence Neu5Ac-alpha-2,3-Gal-beta-1,3-GalNAc. In addition, it is involved in the synthesis of ganglioside GD1A from GM1B. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Transcript variants encoding different isoforms have been found for this gene.
Specifications
| ST6GALNAC4 | |
| Polyclonal | |
| Unconjugated | |
| ST6GALNAC4 | |
| 3-N-acetyl-galactos; alpha-2,6-sialyltransferase ST6GalNAc IV; alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3-N-acetyl-galactosaminide alpha-2,6-sialyltransferase; beta-galactosamide alpha-2,6-sialyltransferase; GalNAc alpha-2,6-sialyltransferase; IV; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2, 6-sialyltransferase alpha2,6-sialyltransferase; neuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase IV; Sia-alpha-2-3-Gal-beta-1-3-GalNAc GalNAc-alpha-2,6-sialyltransferase; sialyltransferase 3C; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) D; sialyltransferase 7D; sialyltransferase 7D ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase); SIAT3C; SIAT3-C; SIAT7B; SIAT7D; SIAT7-D; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 GalNAc alpha-2,6-sialyltransferase 4; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 4; ST6GalNAc; ST6GalNAc IV; ST6GALNAC2; ST6GALNAC4; ST6GalNAcIV; st6GalNAc-IV | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27090 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H4F1 | |
| ST6GALNAC4 | |
| Synthetic peptide directed towards the middle region of human ST6GALNAC4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction