Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ST6GALNAC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531615
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (96%), Pig (93%), Chimpanzee (99%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein prefers glycoproteins rather than glycolipids as substrates and shows restricted substrate specificity, utilizing only the trisaccharide sequence Neu5Ac-alpha-2,3-Gal-beta-1,3-GalNAc. In addition, it is involved in the synthesis of ganglioside GD1A from GM1B. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Transcript variants encoding different isoforms have been found for this gene.
Specifications
ST6GALNAC4 | |
Polyclonal | |
Unconjugated | |
ST6GALNAC4 | |
3-N-acetyl-galactos; alpha-2,6-sialyltransferase ST6GalNAc IV; alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3-N-acetyl-galactosaminide alpha-2,6-sialyltransferase; beta-galactosamide alpha-2,6-sialyltransferase; GalNAc alpha-2,6-sialyltransferase; IV; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2, 6-sialyltransferase alpha2,6-sialyltransferase; neuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase; NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc-alpha-2,6-sialyltransferase IV; Sia-alpha-2-3-Gal-beta-1-3-GalNAc GalNAc-alpha-2,6-sialyltransferase; sialyltransferase 3C; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) D; sialyltransferase 7D; sialyltransferase 7D ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase); SIAT3C; SIAT3-C; SIAT7B; SIAT7D; SIAT7-D; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 GalNAc alpha-2,6-sialyltransferase 4; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 4; ST6GalNAc; ST6GalNAc IV; ST6GALNAC2; ST6GALNAC4; ST6GalNAcIV; st6GalNAc-IV | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27090 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.55 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9H4F1 | |
ST6GALNAC4 | |
Recombinant fragment corresponding to a region within amino acids 83 and 295 of Human ST6GALNAC4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction