Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST6GALNAC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543473 Supplier Invitrogen™ Supplier No. PA543473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AFMITRHKML QFDELFKQET GKDRKISNTW LSTGWFTMTI ALELCDRINV Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

ST6GALNAC5 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions. ST6GALNAC5 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions (Tsuchida et al., 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST6GALNAC5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ST6GALNAC5
Gene Accession No. Q9BVH7
Gene Alias AI851940; Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase V; alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase E; EC 2.4.99; EC 2.4.99.7; galNAc alpha-2,6-sialyltransferase V; GD1 alpha synthase; LOC100715749; MGC3184; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) E; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) E; sialyltransferase 7E; SIAT7E; SIAT7-E; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6 GalNAc alpha-2,6-sialyltransferase 5; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5; ST6GalNAc V; ST6GALNAC5; ST6GalNAcV
Gene Symbols ST6GALNAC5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ST6GALNAC5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81849
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.