Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6GALNAC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 164421AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-0946


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

ST6GALNAC5 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions. ST6GALNAC5 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions (Tsuchida et al., 2003 ).
Specifications

Specifications

ST6GALNAC5
Polyclonal
Unconjugated
ST6GALNAC5
GD1 alpha synthase, Sialyltransferase 7E, SIAT7 E, SIAT7E, ST6GalNAc V, ST6GALNAC5, ST6GalNAcV
Rabbit
Antigen Affinity Chromatography
RUO
26938, 365984, 81849
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BVH7, Q9QYJ1
St6galnac5
ST6GALNAC5 Fusion Protein Ag9767
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.