Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST6GALNAC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553872 Supplier Invitrogen™ Supplier No. PA553872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRCHQCVIVS SSSHLLGTKL GPEIERAECT IRMNDAPTTG YSADVGNKTT YRVVAHSSVF RVLRRPQEFV NRTPETVFIF WGPPSKMQKP QGSLVRVIQR AGLVFPNMEA YAVSPGRMRQ FDDLFRGETG KDREKSHSWL STGWF Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

ST6GALNAC6 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST6GALNAC6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ST6GALNAC6
Gene Accession No. Q969X2
Gene Alias (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1, 3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6; alpha-2,6-sialyltransferase ST6GalNAc VI; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6; beta-galactosamide alpha-2,6-sialyltransferase; CMP-NeuAC:(beta)-N-acetylgalactosaminide (alpha)2,6-sialyltransferase member VI; galNAc alpha-2,6-sialyltransferase VI; Sialyltransferase 7F; sialytransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialytransferase) F; sialytransferase 7F; SIAT7F; SIAT7-F; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6; ST6 GalNAc alpha-2,6-sialyltransferase 6; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6; ST6 -N-acetylgalactosaminide alpha-26-sialyltransferase 6; ST6GalNAc VI; St6galnac6; ST6GalNAcVI; st6GalNAc-VI; UNQ708/PRO1359
Gene Symbols ST6GALNAC6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ST6GALNAC6. Recombinant protein control fragment (Product #RP-101033).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30815
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.