Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558345 Supplier Invitrogen™ Supplier No. PA558345
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: THQFPELMGV FAKAMIDIFC SAEFRDWNCK SIFMRVEDEL EIPPAPQSQH FQ Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 29%.

The gene for this product maps to a region on chromosome 7 identified as an autism-susceptibility locus. Mutation screening of the entire coding region in autistic individuals failed to identify phenotype-specific variants, suggesting that coding mutations for this gene are unlikely to be involved in the etiology of autism. The function of this gene product has not been determined. Transcript variants encoding different isoforms of this protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene St7
Gene Accession No. Q9NRC1
Gene Alias 4low density lipoprotein-related protein 12; 9430001H04Rik; ETS7q; FAM4A; FAM4A1; Fam4a2; family with sequence similarity 4, subfamily A, member 1; family with sequence similarity 4, subfamily A, member 2; HELG; mRay; Protein FAM4A1; Protein HELG; RAY1; SEN4; St7; suppression of tumorigenicity 7; suppression of tumorigenicity 7 (breast); suppressor of tumorigenicity 7 protein; TSG7
Gene Symbols St7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ST7. Recombinant protein control fragment (Product #RP-98322).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7982
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.