Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA558344
Description
Immunogen sequence: PQARISAAHE ALEINEIRSR VEVPLIASST IWEIKLLPKC ATAY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The gene for this product maps to a region on chromosome 7 identified as an autism-susceptibility locus. Mutation screening of the entire coding region in autistic individuals failed to identify phenotype-specific variants, suggesting that coding mutations for this gene are unlikely to be involved in the etiology of autism. The function of this gene product has not been determined. Transcript variants encoding different isoforms of this protein have been described.
Specifications
ST7 | |
Polyclonal | |
Unconjugated | |
ST7 | |
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7, family with sequence similarity 4, subfamily A, member 1, suppression of tumorigenicity 7 (breast), suppressor of tumorigenicity 7 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7982 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRC1 | |
ST7 | |
Recombit protein corresponding to Human ST7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction