Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST7L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559948 Supplier Invitrogen™ Supplier No. PA559948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADRGGVGEA AAVGASPASV PGLNPTLGWR ERLRAGL Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 60%.

This gene was identified by its similarity to the ST7 tumor suppressor gene found in the chromosome 7q31 region. This gene is clustered in a tail-to-tail manner with the WNT2B gene in a chromosomal region known to be deleted and rearranged in a variety of cancers. Several transcript variants encoding many different isoforms have been described, but some have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST7L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene St7l
Gene Accession No. Q8TDW4
Gene Alias FAM4B; ST7L; ST7R; ST7-related protein; STLR; suppression of tumorigenicity 7 like; suppression of tumorigenicity 7-like; suppressor of tumorigenicity 7 protein-like; suppressor of tumorigenicity protein 7-like protein; tumor suppressor ST7 like
Gene Symbols St7l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ST7L. Recombinant protein control fragment (Product #RP-90913).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54879
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.