Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST8SIA1, Mouse anti-Human, Clone: HIC0-3C5, Millipore Sigma™

Catalog No. MABS199725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS199725 25 μg
MABS1997100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS199725 Supplier MilliporeSigma Supplier No. MABS199725UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-ST8SIA1, clone HIC0-3C5, Cat. No. MABS1997, is a mouse monoclonal antibody that detects Alpha-N-acetylneuraminide alpha-2,8-sialyltransferase and has been tested for use in Flow Cytometry, Immunoflurescence, and Immunohistochemistry.

Alpha-N-acetylneuraminide alpha-2,8-sialyltransferase (UniProt: Q92185; also known as EC: 2.4.99.8, Alpha-2,8-sialyltransferase 8A, Ganglioside GD3 synthase, Ganglioside GT3 synthase, Sialyltransferase 8A, SIAT8-A, Sialyltransferase St8Sia I, ST8SiaI) I encoded by the ST8SIA1 (also known as SIAT8, SIAT8A) gene (Gene ID: 6489) in human. SIAT8-A is a single-pass type II membrane protein that is strongly expressed in melanoma cell lines, adult and fetal brain, and to a lesser extent in adult and fetal lung. It is involved in the production of gangliosides GD3 and GT3 from GM3. It catalyzes the transfer of sialic acid from CMP-sialic acid to GM3 to produce GD3 and GT3 in a successive manner. It is also involved in the pathway protein glycosylation. SIAT8-A contains a cytoplasmic domain (aa 1-29); a transmembrane domain (aa 30-48); and a lumenal domain (aa 49-356). Two isoforms of SIAT8-A have been described that are produced by alternative splicing. (Ref.: Dorrell, C., et al. (2016). Nat. Commun. 7; 11756).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST8SIA1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone HIC0-3C5
Conjugate Unconjugated
Dilution Flow Cytometry Analysis: A representative lot detected ST8SIA1 in Flow Cytometry applications (Dorrell, C., et. al. (2016). Nat Commun. 7:11756).Immunofluorescence Analysis: A representative lot detected ST8SIA1 in Immunofluorescence applications (Dorrell, C., et. al. (2016). Nat Commun. 7:11756).
Formulation Purified mouse monoclonal antibody IgM in PBS with 0.05% sodium azide.
Gene Alias Alpha-N-acetylneuraminide alpha-2;8-sialyltransferase;EC: 2.4.99.8;Alpha-2;8-sialyltransferase 8A;Ganglioside GD3 synthase;Ganglioside GT3 synthase;Sialyltransferase 8A;SIAT8-A;Sialyltransferase St8Sia I;ST8SiaI
Gene Symbols ST8SIA1;SIAT8;SIAT8A
Host Species Mouse
Immunogen Trypsin-dissociated human pancreatic islet cells with low levels of exocrine and ductal cells.
Purification Method Purified by ion-exchange chromatography
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_003025.1
Test Specificity Clone HIC0-3C5 specifically detects human Alpha-N-acetylneuraminide alpha-2,8-sialyltransferase (ST8SIA1).
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.