Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ST8SIA2 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547856
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
The protein encoded by this gene is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. The encoded protein may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29.
Specifications
ST8SIA2 | |
Polyclonal | |
Unconjugated | |
ST8SIA2 | |
AI323367; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 8B; alpha-2,8-sialyltransferase 8B 1; cb394; HsT19690; id:ibd5161; MGC116854; MGC116857; N-glycan alpha 2,8-sialyltransferase; polysialic acid synthase; sialyltransferase 8; sialyltransferase 8 (alpha-2 8-sialytransferase) B; sialyltransferase 8 (alpha-2, 8-sialyltransferase) B; sialyltransferase 8 (alpha-2, 8-sialytransferase) B; sialyltransferase 8B; Sialyltransferase St8Sia II; sialyltransferase X; sialytransferase St8Sia II; SIAT 8B; siat8; SIAT8B; SIAT8-B; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 2; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2; ST8Sia II; ST8SIA2; st8sia2 protein; ST8SiaII; ST8SIA-II; STX | |
Sheep | |
Antigen Affinity Chromatography | |
RUO | |
8128 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q92186 | |
ST8SIA2 | |
Mouse myeloma cell line NS0-derived recombinant human alpha-2,8-Sialyltransferase8B/ST8SIA2 Asp24-Thr375. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction