Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST8SIA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113279
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113279 Supplier Invitrogen™ Supplier No. PA5113279
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ST8SIA2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The protein encoded by this gene is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. The encoded protein may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST8SIA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ST8SIA2
Gene Accession No. O35696, Q92186
Gene Alias AI323367; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 8B; alpha-2,8-sialyltransferase 8B 1; cb394; HsT19690; id:ibd5161; MGC116854; MGC116857; N-glycan alpha 2,8-sialyltransferase; polysialic acid synthase; sialyltransferase 8; sialyltransferase 8 (alpha-2 8-sialytransferase) B; sialyltransferase 8 (alpha-2, 8-sialyltransferase) B; sialyltransferase 8 (alpha-2, 8-sialytransferase) B; sialyltransferase 8B; Sialyltransferase St8Sia II; sialyltransferase X; sialytransferase St8Sia II; SIAT 8B; siat8; SIAT8B; SIAT8-B; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 2; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2; ST8Sia II; ST8SIA2; st8sia2 protein; ST8SiaII; ST8SIA-II; STX
Gene Symbols ST8SIA2
Host Species Rabbit
Immunogen Recombinant Human Alpha-2,8-sialyltransferase 8B protein (24-160AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20450, 8128
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.