Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST8SIA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569551 Supplier Invitrogen™ Supplier No. PA569551
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

The protein encoded by this gene catalyzes the polycondensation of alpha-2,8-linked sialic acid required for the synthesis of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). The encoded protein, which is a member of glycosyltransferase family 29, is a type II membrane protein that may be present in the Golgi apparatus. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST8SIA4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene St8sia4
Gene Accession No. 0
Gene Alias alpha-2,8-polysialyltransferase; alpha-2,8-polysialyltransferase IV; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 8D; alpha-2,8-sialyltransferase ST8Sia IV; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyl transferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase-like protein; LOW QUALITY PROTEIN: CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase; N-glycan alpha 2,8-sialyltransferase; polysialyltransferase; polysialyltransferase-1; Pst; PST1; PST-1; Sia8D; sialyltransf; sialyltransferase 8 (alpha-2 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2, 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-sialyltransferase) D; sialyltransferase 8 (alpha-2, 8-sialytransferase) D; sialyltransferase 8D; Sialyltransferase St8Sia IV; sialytransferase St8Sia IV; siat 8D; SIAT8D; SIAT8-D; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 4; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4; ST8Sia IV; St8sia4; st8sia4 protein; ST8SiaIV; ST8SIA-IV
Gene Symbols St8sia4
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat ST8SIA4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116696
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.