Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST8SIA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526774
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526774 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526774 Supplier Invitrogen™ Supplier No. PA526774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology.

The protein encoded by this gene catalyzes the polycondensation of alpha-2,8-linked sialic acid required for the synthesis of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). The encoded protein, which is a member of glycosyltransferase family 29, is a type II membrane protein that may be present in the Golgi apparatus. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST8SIA4
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene St8sia4
Gene Accession No. Q64692, Q92187
Gene Alias alpha-2,8-polysialyltransferase; alpha-2,8-polysialyltransferase IV; alpha-2,8-sialyltransferase; alpha-2,8-sialyltransferase 8D; alpha-2,8-sialyltransferase ST8Sia IV; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyl transferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase; CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase-like protein; LOW QUALITY PROTEIN: CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase; N-glycan alpha 2,8-sialyltransferase; polysialyltransferase; polysialyltransferase-1; Pst; PST1; PST-1; Sia8D; sialyltransf; sialyltransferase 8 (alpha-2 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-polysialyltransferase) D; sialyltransferase 8 (alpha-2, 8-polysialytransferase) D; sialyltransferase 8 (alpha-2, 8-sialyltransferase) D; sialyltransferase 8 (alpha-2, 8-sialytransferase) D; sialyltransferase 8D; Sialyltransferase St8Sia IV; sialytransferase St8Sia IV; siat 8D; SIAT8D; SIAT8-D; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 4; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4; ST8Sia IV; St8sia4; st8sia4 protein; ST8SiaIV; ST8SIA-IV
Gene Symbols St8sia4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 186-214 amino acids from the central region of human ST8SIA4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20452, 7903
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.