Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ST8SIA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552845 Supplier Invitrogen™ Supplier No. PA552845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RILVEESREA THGTPAALRT LRSPATAVPR ATNSTYLNEK SLQLTEKCKN LQYGIESFSN KTKGYSENDY LQIITDIQSC PWKRQAEEYA NFRAKLASCC DAVQNFVVSQ NNTPVGTNMS YEVESKKEIP IKKNIFHMFP VSQP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 48%.

Prefers O-glycans to N-glycans or glycolipids as acceptor substrates. The minimal acceptor substrate is the NeuAc- alpha-2,3(6)-Gal sequence at the non-reducing end of their carbohydrate groups.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST8SIA6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ST8SIA6
Gene Accession No. P61647
Gene Alias Alpha-2,8-sialyltransferase 8F; alpha-2,8-sialyltransferase 8F variant 1; alpha-2,8-sialyltransferase 8F variant 2; alpha-2,8-sialyltransferase 8F variant 3; SIA8F; Sialyltransferase 8F; sialyltransferase 8F (alpha-2, 8-sialyltransferase); sialyltransferase St8Sia VI; sialytransferase St8Sia VI; SIAT8F; SIAT8-F; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 6; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6; ST8SIA6; ST8SiaVI; ST8SIA-VI
Gene Symbols ST8SIA6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ST8SIA6. Recombinant protein control fragment (Product #RP-90326).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338596
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.