Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAB2 Monoclonal Antibody (OTI2E12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA806361

Description
Phosphatidylserine receptor that enhances the engulfment of apoptotic cells. Hyaluro receptor that binds to and mediates endocytosis of hyaluronic acid (HA). Acts also, in different species, as a primary systemic scavenger receptor for heparin (Hep), chondroitin sulfate (CS), dermatan sulfate (DS), nonglycosaminoglycan (GAG), acetylated low-density lipoprotein (AcLDL), pro-collagen propeptides and advanced glycation end products (AGE). May serve to maintain tissue integrity by supporting extracellular matrix turnover or it may contribute to maintaining fluidity of bodily liquids by resorption of hyaluro. Counter receptor which plays an important role in lymphocyte recruitment in the hepatic vasculature. Binds to both Gram-positive and Gram-negative bacteria and may play a role in defense against bacterial infection. The proteolytically processed 190 kDa form also functions as an endocytosis receptor for heparin internalization as well as HA and CS.Specifications
STAB2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WWQ8 | |
STAB2 | |
Human recombit protein fragment corresponding to amino acids 1914-2194 of human STAB2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI2E12 | |
Unconjugated | |
STAB2 | |
FEEL2, FELE-2, FELL2, FEX2, HARE, SCARH1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55576 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction