Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stabilin-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
| Antigen | Stabilin-2 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Stabilin-2 Polyclonal specifically detects Stabilin-2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Stabilin-2 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CD44-like precursor FELL, DKFZP434E0321, FAS1 EGF-like and X-link domain containing adhesion molecule-2, FAS1 EGF-like and X-link domain-containing adhesion molecule 2, fasciclin egf-like, laminin-type egf-like, and link domain-containing scavengerreceptor-2, Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavengerreceptor 2, FEEL2, FEEL-2, FELE-2, FELL, FEX2, HAREFELL2, hepatic hyaluronan clearance receptor, Hyaluronan receptor for endocytosis, hyaluronic acid receptor for endocytosis, STAB-2, stabilin 2, stabilin-2 | |
| STAB2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8WWQ8 | |
| 55576 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VVLEEKLLKNDLHNGMHRETMLGFSYFLSFFLHNDQLYVNEAPINYTNVATDKGVIHGLGKVLEIQKNRCDNNDTTIIRGRCRTCSSELTCPFGTKSLGNEKRRCIYTSYFMGRRTLFIGCQPKCVRTVITR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title