Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stabilin 2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Supplier: Thermo Scientific PA547522
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human Stabilin-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Stabilin 2 Polyclonal antibody specifically detects Stabilin 2 in Human samples. It is validated for Flow Cytometry, Western Blot.
Specifications
Stabilin 2 | |
Polyclonal | |
Unconjugated | |
STAB2 | |
190 kDa form stabilin-2; 190 kDa hyaluronan receptor for endocytosis; CD44-like precursor FELL; DKFZP434E0321; FAS1 EGF-like and X-link domain containing adhesion molecule-2; FAS1 EGF-like and X-link domain-containing adhesion molecule 2; fasciclin egf-like, laminin-type egf-like, and link domain-containing scavenger receptor-2; fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavenger receptor 2; FEEL2; FEEL-2; FELE-2; FELL; FELL2; FELL-2; FEX2; Hare; HAREFELL2; hepatic hyaluronan clearance receptor; hyaluronan receptor for endocytosis; hyaluronic acid receptor for endocytosis; MFEEL-2; SCARH1; scavenger receptor FEEL-2; Short form stabilin-2; stab2; STAB-2; stab2 {ECO:0000250; stabilin 2; Stabilin2; stabilin-2; UniProtKB:Q8WWQ8} | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
55576 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8WWQ8 | |
STAB2 | |
E. coli-derived recombinant human Stabilin-2 Gly2198-Val2295 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction