Learn More
Thermo Scientific™ Heratherm Stability Chamber
Description
- Broad Relative Humidity range to cover many applications, including Humidity levels below ambient.
- Environmentally friendly water-efficient humidification system.
- Minimal condensate in standard applications.
- Integrated water tank; fixed water connection for in-house purified water option available.
- Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
- Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
- Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
- Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
- Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
- Simplified setup with no technician required; install using standard power cord.
- IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
- Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
- Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
- Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
Specifications
Specifications
| Alarm Types | Audible and Visible |
| Capacity (English) | 26.5 cu. ft. |
| Capacity (English) Shelf | 66.1 lb. |
| Capacity (Metric) Shelf | 30 kg |
| Certifications/Compliance | UL, CE |
| Data Outputs | USB, Ethernet |
| Capacity (Metric) | 750 L |
| Chamber Material | Powder-coated Cold Rolled Steel |
| Material | Powder-coated Cold Rolled Steel |
| No. of Shelves | 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.