Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557175
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557175 Supplier Invitrogen™ Supplier No. PA557175
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSDSPHRTST SDLVEVPEEA NGPGGGYDLR KRSNSVFTYP ENGTDDFRDP AKNINHQGSL SKDPLQMNTY Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 83%.

Promotes expression of the ion channel CACNA1H at the cell membrane, and thereby contributes to the regulation of channel activity. Plays a minor and redundant role in promoting the expression of calcium channel CACNA1S at the cell membrane, and thereby contributes to increased channel activity. Slows down the inactivation rate of the calcium channel CACNA1C. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STAC
Gene Accession No. Q99469
Gene Alias SH3 and cysteine rich domain; SH3 and cysteine-rich domain-containing protein; Src homology 3 and cysteine-rich domain-containing protein; src homology three (SH3) and cysteine rich domain; Stac; STAC1
Gene Symbols STAC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STAC. Recombinant protein control fragment (Product #RP-96355).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6769
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.