Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598809
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598809 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598809 Supplier Invitrogen™ Supplier No. PA598809
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STAC Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Promotes expression of the ion channel CACNA1H at the cell membrane, and thereby contributes to the regulation of channel activity. Plays a minor and redundant role in promoting the expression of calcium channel CACNA1S at the cell membrane, and thereby contributes to increased channel activity. Slows down the inactivation rate of the calcium channel CACNA1C. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAC
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene STAC
Gene Accession No. P97306, Q99469
Gene Alias SH3 and cysteine rich domain; SH3 and cysteine-rich domain-containing protein; Src homology 3 and cysteine-rich domain-containing protein; src homology three (SH3) and cysteine rich domain; Stac; STAC1
Gene Symbols STAC
Host Species Rabbit
Immunogen Recombinant Human SH3 and cysteine-rich domain-containing protein (1-119AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20840, 6769
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.