Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101884 Supplier Invitrogen™ Supplier No. PA5101884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total STAC2.

STAC2 (SH3 and cysteine-rich domain-containing protein 2), also known as 24b2/STAC2, is a 411 amino acid protein that contains one SH3 domain and one phorbol-ester/DAG-type zinc finger. Encoding two alternatively spliced isoforms, the STAC2 gene contains over 15 thousand bases and maps to human chromosome 17q12. Human chromosome 17 comprises over 2.5% of the human genome and encodes over 1, 200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene STAC2
Gene Accession No. Q6ZMT1, Q8R1B0
Gene Alias 24b2; 24b2/STAC2; AW240854; SH3 and cysteine rich domain 2; SH3 and cysteine-rich domain-containing protein 2; Src homology 3 and cysteine-rich domain-containing protein 2; Stac2
Gene Symbols STAC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human STAC2(Accession Q6ZMT1), corresponding to amino acid residues A215-P265.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217154, 342667, 363674
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.