Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAG1 Monoclonal Antibody (GT1515)
GREENER_CHOICE

Catalog No. PIMA531524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531524 Supplier Invitrogen™ Supplier No. MA531524
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%). Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the SCC3 family and is expressed in the nucleus. It encodes a component of cohesin, a multisubunit protein complex that provides sister chromatid cohesion along the length of a chromosome from DNA replication through prophase and prometaphase, after which it is dissociated in preparation for segregation during anaphase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAG1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT1515
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene STAG1
Gene Accession No. Q8WVM7
Gene Alias AU045003; cohesin subunit SA-1; DKFZp781D1416; nuclear protein stromal antigen 1; SA1; SA-1; SCC3; SCC3 homolog 1; SCC3A; STAG1; Stromal antigen 1
Gene Symbols STAG1
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SA1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10274, 315958
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.