Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115861 Supplier Invitrogen™ Supplier No. PA5115861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SA1.

This gene is a member of the SCC3 family and is expressed in the nucleus. It encodes a component of cohesin, a multisubunit protein complex that provides sister chromatid cohesion along the length of a chromosome from DNA replication through prophase and prometaphase, after which it is dissociated in preparation for segregation during anaphase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene STAG1
Gene Accession No. Q8WVM7, Q9D3E6
Gene Alias AU045003; cohesin subunit SA-1; DKFZp781D1416; nuclear protein stromal antigen 1; SA1; SA-1; SCC3; SCC3 homolog 1; SCC3A; STAG1; Stromal antigen 1
Gene Symbols STAG1
Host Species Rabbit
Immunogen A synthesized peptide derived from human STAG1(Accession Q8WVM7), corresponding to amino acid residues S45-K95.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10274, 20842, 315958
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.