Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAG1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993499
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3499 10 μL
50-199-3498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3499 Supplier Active Motif Supplier No. 61562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the SCC3 family and is expressed in the nucleus. It encodes a component of cohesin, a multisubunit protein complex that provides sister chromatid cohesion along the length of a chromosome from DNA replication through prophase and prometaphase, after which it is dissociated in preparation for segregation during anaphase.

Specifications

Antigen STAG1
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene STAG1
Gene Accession No. Q8WVM7
Gene Alias AU045003; Cohesin subunit SA-1; DKFZp781D1416; nuclear protein stromal antigen 1; SA1; SA-1; SCC3; SCC3 homolog 1; SCC3A; Stag1; Stromal antigen 1
Gene Symbols STAG1
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the C-terminal region of human STAG1.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10274
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.