Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAG2 Antibody, Allophycocyanin, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100300APC
Description
STAG2 Polyclonal antibody specifically detects STAG2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
STAG2 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
bA517O1.1, cohesin subunit SA-2, DKFZp686P168, SA-2, SA2FLJ25871, SCC3 homolog 2, stromal antigen 2DKFZp781H1753 | |
A 31 amino acid synthetic peptide, which represented a portion of human Stromal Antigen 2 encoded within exon 31 (LocusLink ID 10735). Host: Goat A 31 amino acid synthetic peptide, which represented a portion of human Stromal Antigen 2 encoded within exon 31 (LocusLink ID 10735). | |
0.1 mL | |
DNA Repair | |
10735 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
APC | |
PBS | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction