Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAG2 Antibody (SR1161), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321847100UL
Description
STAG2 Monoclonal antibody specifically detects STAG2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
STAG2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR1161 | |
Western Blot 1:500-1:1000, Flow Cytometry 1:200, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
bA517O1.1, cohesin subunit SA-2, DKFZp686P168, SA-2, SA2FLJ25871, SCC3 homolog 2, stromal antigen 2DKFZp781H1753 | |
A synthesized peptide derived from human STAG2 (Uniprot #: Q8N3U4) | |
100 μg | |
DNA Repair | |
10735 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction