Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAG2 Monoclonal Antibody (GT1417)
GREENER_CHOICE

Catalog No. PIMA531532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531532 Supplier Invitrogen™ Supplier No. MA531532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Rabbit (100%), Bovine (100%). Positive Control: 293T, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

STAG2 is a component of the cohesin complex. This complex is required for the cohesion of sister chromatids after DNA replication. The cohesin complex forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The cohesin complex may also play a role in spindle pole assembly during mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAG2
Applications Western Blot
Classification Monoclonal
Clone GT1417
Concentration 2.39 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene STAG2
Gene Accession No. Q8N3U4
Gene Alias 9230105L23Rik; B230112I07Rik; bA517O1.1; cohesin subunit SA-2; DKFZp686P168; DKFZp781H1753; FLJ25871; nuclear protein SA2; OTTHUMP00000024338; OTTHUMP00000043514; OTTHUMP00000043515; OTTHUMP00000081754; OTTHUMP00000081755; RGD1562042; RP11-517O1.1; SA2; SA-2; Sap2; SCC3 homolog 2; SCC3B; STAG2; Stromal antigen 2
Gene Symbols STAG2
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SA2. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10735
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.