Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STAG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578339
Description
Positive Control: 293T, A431, HeLa, HepG2, Neuro2A, CD830, NIH-3T3, Raw264.7, C2C12 Predicted Reactivity: Rat (99%), Xenopus laevis (81%), Rabbit (98%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STAG2 is a component of the cohesin complex. This complex is required for the cohesion of sister chromatids after DNA replication. The cohesin complex forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The cohesin complex may also play a role in spindle pole assembly during mitosis.
Specifications
STAG2 | |
Polyclonal | |
Unconjugated | |
STAG2 | |
9230105L23Rik; B230112I07Rik; bA517O1.1; cohesin subunit SA-2; DKFZp686P168; DKFZp781H1753; FLJ25871; nuclear protein SA2; OTTHUMP00000024338; OTTHUMP00000043514; OTTHUMP00000043515; OTTHUMP00000081754; OTTHUMP00000081755; RGD1562042; RP11-517O1.1; SA2; SA-2; Sap2; SCC3 homolog 2; SCC3B; STAG2; Stromal antigen 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10735, 20843 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O35638, Q8N3U4 | |
STAG2 | |
Recombinant protein encompassing a sequence within the N-terminus region of human SA2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction