Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563556 Supplier Invitrogen™ Supplier No. PA563556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LFHQDKQLLL SYLEKCLQHV SQAPGHPWGP VTTYCHSLSP VENTAETSPQ VLPSSKRRRV EGPAKPNRED VSS Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 71%.

STAG3 is a meiosis specific component of cohesin complex. The cohesin complex is required for the cohesion of sister chromatids after DNA replication. The cohesin complex apparently forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The meiosis-specific cohesin complex probably replaces mitosis specific cohesin complex when it dissociates from chromatin during prophase I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAG3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STAG3
Gene Accession No. Q9UJ98
Gene Alias cohesin subunit SA-3; MGC119262; MGC119264; MGC119265; NHLRC 1; SA-2; SCC3 homolog 3; Stag3; Stromal antigen 3; stromalin 3; stromalin-3
Gene Symbols STAG3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STAG3. Recombinant protein control fragment (Product #RP-103326).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.