Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Stainless Steel Desiccator with Stainless Steel Shelves
GREENER_CHOICESDPSCI_ED

Catalog No. p-4006914 Shop All Fisherbrand Products
Click to view available options
Size:
Large
Standard

Catalog No. S02123A

Only null left
Add to Cart
Add to Cart
Request a Quote

Includes

Two stainless-steel shelves, desiccant tray

One piece neoprene gasket provides excellent sealing of the chamber.

Key Features

  • Polished stainless steel with 0.187 in. thick (4.8mm) glass
  • Airtight door with neoprene gasket, full-height hinges and double-action bar locks
  • Fixed wire shelf brackets every 0.5 in. (13mm)

Standard Size

  • Glass on door and sides
  • Tray holds 16 oz. (454g) of desiccant
  • Weighs 23 lb. (10kg)

Large Size

  • Glass on door and back, stainless-steel sides
  • Shelf dimensions: 13.375 x 18 in. (34 x 46cm)
  • Weighs 57 lb. (26kg)

Warranty and Services

Two years

Specifications

Material Stainless Steel
Product Type Desiccator Cabinet
For Use With (Application) Storing reagents or soil samples, drying biological specimens or storage sludge samples
Includes Airtight door with neoprene gasket, full-height hinges, double-action bar locks, two stainless steel shelves and desiccant tray
Size Large
Length (English) 22.5 in.
Length (Metric) 56.5 cm
Width (English) 16.5 in.
Width (Metric) 41.9 cm
Height (English) 23 in.
Height (Metric) 58 cm
Weight (English) 57 lb.
Weight (Metric) 26 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.