Learn More
Stainless Steel foil, 0.5mm (0.02in) thick, Type 304
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Silver |
| Chemical Name or Material | Stainless Steel |
| Appearance | Silver -colored foil |
| Quantity | 25 x 25 mm |
| Odor | Odorless |
| Form | Foil |
| Molecular Formula | Fe:Cr:Ni; 70:19:11 wt% |
| MDL Number | MFCD00148499 |
| Solubility Information | It is insoluble in water. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.