Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Stainless Steel Forceps, Economy Blunt
SDPSCI_ED

Catalog No. S01329 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (English):
4.5 in.
5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S01329 5 in.
S01328 4.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S01329 Supplier United Scientific™ Supplier No. FOBL05
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economy Blunt Stainless Steel Forceps ideal for general use in laboratories and classrooms.

  • Good quality stainless steel forceps for general use in the laboratory.
  • All feature serrated tips.
  • Non-sterile.
  • Autoclavable.

Specifications

Length (English) 5 in.
Length (Metric) 12.7 cm
Material Stainless Steel
Style Standard
Tip Style Blunt
Product Type Forceps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.