Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 304

Catalog No. AA13476CH
Click to view available options
Quantity:
10 x 10 cm
20 x 30 cm
30 x 60 cm

Fe:Cr:Ni; 70:19:11 wt.%

It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stainless Steel gauze
Name Note Open area: 36%; Width of opening: 0.003 in.
Quantity 20 x 30 cm
Mesh Size 200 Mesh
Form Wire Cloth
Molecular Formula Fe:Cr:Ni; 70:19:11 wt.%
MDL Number MFCD00148499
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.