Learn More
RETSCH Stainless Steel Grinding Jar
Catalog No. 0841759 Shop All RETSCH Inc. Products
Change view
Click to view available options
For Use With (Equipment):
For MM 200
Capacity (Metric):
10 mL
Catalog No. For Use With (Equipment) Capacity (Metric)
08-417-59 For MM 200 10 mL
1 options
Catalog No. 08-417-59 Supplier RETSCH Supplier No. 024620061
Add to Cart
Add to Cart
Use these grinding jars for dry, wet, and cryogenic grinding of small samples. Retsch™ Stainless Steel Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
  • Screw-top grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Product Type Grinding Jar
Capacity (Metric) 10 mL
For Use With (Equipment) For MM 200
Material Stainless Steel