Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Stainless Steel Spatulas, Flat/Tapered
SDPSCI_ED

Catalog No. S15819 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric):
6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S15819 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15819 Supplier United Scientific™ Supplier No. SSFT06
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economical option for everyday lab use.

  • Pack of 10 stainless steel laboratory spatulas.
  • One end flat, one end tapered.
  • 6 inches in length.

Specifications

Quantity 1 Ea.
Blade Material Stainless Steel
Handle Material Stainless Steel
Material Stainless Steel
Handle Type No Handle
Length (Metric) 6 in.
Autoclavable Autoclavable
Shape Tapered Flat End
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.