Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Stains-All, 96%

Catalog No. p-7082206
Click to view available options
Quantity:
1g
5g

CAS: 7423-31-6 | C30H27BrN2S2

General protein and nucleic acid stain, which stains different colors on bands of RNA (bluish purple), DNA (blue) and proteins (red)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-31-6
Molecular Formula C30H27BrN2S2
Synonym 1-Ethyl-2-[(E)-3-(1-ethylnaphtho[1,2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1,2-d]thiazolium bromide; 3,3′-Diethyl-9-methyl-4,5,4′,5′-dibenzothiacarbocyanine

Specifications

Formula Weight 559.58g/mol
Beilstein 3865213
Product Type Stain
Quantity 1g
Assay 0.96
EINECSNumber 231-047-7
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.