Learn More
Stak-a-Cab™ Stackable Cabinets
Shop All SciMatCo Products

Description
Includes
Each part number represent one side-by-side cabinet; set of three cabinets shown represents three separate items stacked with base.
No interior metal parts to corrode: wooden hinges and handles, polypropylene lining on cabinet floor, and polypropylene padlock assembly.
- Lock with two keys and piano-style door hinges
- All hardware is corrosion resistant
- Liquid-tight trough floors
- 10 x 10 in. (25 x 25cm) Hazard labels on both doors
- Epoxy paint finish inside and out
Corrosives Cabinet
- Capacity: 22 x 2.5L (approx. 14.5 gal./55L)
- Blue finish
- Accepts nitric acid compartment
Flammables Cabinet
- Capacity: 16 x 1 gal. (approx. 60.5L)
- Yellow finish
Combination Cabinet
- Combination corrosives/flammables cabinet
- Corrosives compartment capacity: 10 x 2.5L (approx. 6.5 gal./25L)
- Flammables compartment capacity: 8 x 1 gal. (approx. 30L)
- Gray finish
Floor Stands
- Height: 4.5 in. (11cm)
- Epoxy paint finish
Order Info
Interior: 15.125D x 45W x 16 in.H (40 x 114 x 41cm); Exterior: 18L x 47W x 18 in.H (46 x 119 x 46cm)
Compliance
Meet all applicable OSHA and NFPA standards.
Specifications
Specifications
| Dimensions (D x W x H) | 18 x 47 x 18 in. |
| Door Style | Manual-close |
| No. of Doors | 2 |
| Surface Coating | Epoxy |
| Depth (English) Exterior | 18 in. |
| Height (Metric) Exterior | 45.7 cm |
| Load Bearing Capacity | 22 x 2.5 L |
| Product Type | Storage Cabinet |
| Width (English) Exterior | 47 in. |
| Includes | Wooden hinges and handles, Polypropylene lining on cabinet floor and Polypropylene padlock assembly, Plastic hasp and padlock, 2 in. liquid tight trough |
| Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.