Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stak-a-Cab™ Stackable Cabinets
SCI_ED

Catalog No. p-2378469 Shop All SciMatCo Products
Encompass
Click to view available options
Color:
Blue
Gray
Yellow
Product Type:
Floor Stand
Storage Cabinet

Includes

Each part number represent one side-by-side cabinet; set of three cabinets shown represents three separate items stacked with base.

Use individually or stack up to three high with the optional floor stand.

No interior metal parts to corrode: wooden hinges and handles, polypropylene lining on cabinet floor, and polypropylene padlock assembly.

  • Lock with two keys and piano-style door hinges
  • All hardware is corrosion resistant
  • Liquid-tight trough floors
  • 10 x 10 in. (25 x 25cm) Hazard labels on both doors
  • Epoxy paint finish inside and out

Corrosives Cabinet

  • Capacity: 22 x 2.5L (approx. 14.5 gal./55L)
  • Blue finish
  • Accepts nitric acid compartment

Flammables Cabinet

  • Capacity: 16 x 1 gal. (approx. 60.5L)
  • Yellow finish

Combination Cabinet

  • Combination corrosives/flammables cabinet
  • Corrosives compartment capacity: 10 x 2.5L (approx. 6.5 gal./25L)
  • Flammables compartment capacity: 8 x 1 gal. (approx. 30L)
  • Gray finish

Floor Stands

  • Height: 4.5 in. (11cm)
  • Epoxy paint finish

Order Info

Interior: 15.125D x 45W x 16 in.H (40 x 114 x 41cm); Exterior: 18L x 47W x 18 in.H (46 x 119 x 46cm)

Compliance

Meet all applicable OSHA and NFPA standards.

Specifications

Dimensions (D x W x H) 18 x 47 x 18 in.
Door Style Manual-close
No. of Doors 2
Surface Coating Epoxy
Depth (English) Exterior 18 in.
Height (Metric) Exterior 45.7 cm
Load Bearing Capacity 22 x 2.5 L
Product Type Storage Cabinet
Width (English) Exterior 47 in.
Includes Wooden hinges and handles, Polypropylene lining on cabinet floor and Polypropylene padlock assembly, Plastic hasp and padlock, 2 in. liquid tight trough
Width (Metric) Exterior 119.4 cm
Color Blue
Height (English) Exterior 18 in.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.