Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAM-1 Antibody (29C678), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056533B
Description
STAM-1 Monoclonal specifically detects STAM-1 in Human samples. It is validated for Western Blot.Specifications
STAM-1 | |
Monoclonal | |
Biotin | |
Q92783 | |
STAM | |
This antibody was developed against a synthetic peptide corresponding to amino acids 149-165 of human STAM. | |
0.1 mL | |
Primary | |
A 59 kDa band is observed. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
29C678 | |
Western Blot | |
DKFZp686J2352, HSE1 homolog, signal transducing adapter molecule 1, signal transducing adaptor molecule (SH3 domain and ITAM motif) 1, STAM1STAM-1 | |
Mouse | |
Protein G purified | |
RUO | |
8027 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction