Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAM-1 Antibody (29C678), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | STAM-1 |
---|---|
Clone | 29C678 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-2 ug/ml |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056533T
![]() |
Novus Biologicals
NB10056533SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056533
![]() |
Novus Biologicals
NB10056533 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
STAM-1 Monoclonal specifically detects STAM-1 in Human samples. It is validated for Western Blot.Specifications
STAM-1 | |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
DKFZp686J2352, HSE1 homolog, signal transducing adapter molecule 1, signal transducing adaptor molecule (SH3 domain and ITAM motif) 1, STAM1STAM-1 | |
STAM | |
IgG1 | |
Protein G purified | |
A 59 kDa band is observed. |
29C678 | |
Western Blot 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Q92783 | |
8027 | |
This antibody was developed against a synthetic peptide corresponding to amino acids 149-165 of human STAM. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title