Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAM Recombinant Rabbit Monoclonal Antibody (HL1635)
GREENER_CHOICE

Catalog No. FEMA550325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550325 Supplier Invitrogen™ Supplier No. MA550325
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

STAM Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

STAM (Signal transducing adaptor molecule) is an adaptor molecule involved in downstream signaling pathways mediated by growth factors and cytokines. STAM also plays an important role in endocytosis, the JAK-STAT signaling pathway, and membrane trafficking. In humans, the gene encoding STAM is located on chromosome 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAM
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1635
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene STAM
Gene Accession No. P70297, Q92783
Gene Alias DKFZp686J2352; HSE1 homolog; RGD1564499; signal transducing adapter molecule 1; signal transducing adaptor molecule; signal transducing adaptor molecule (SH3 domain and ITAM motif) 1; Stam; STAM1; STAM-1; STAT91
Gene Symbols STAM
Host Species Rabbit
Immunogen Recombinant fragment of human STAM.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20844, 8027
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.