Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121061 Supplier Invitrogen™ Supplier No. PA5121061
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: MCF7, HepG2, OVCAR3, PC-3. The target is usually found in the following locations: Cytoplasm, Cytoplasmic side, Early endosome membrane, Peripheral membrane protein. Immunogen sequence: KLHPPAHYPP ASSGVPMQTY PVQSHGGNYM GQSIHQVTVA QSYSLGPDQI GPLRSLPPNV NSSVTAQPAQ TSYLSTGQDT VSNPTYMNQN SNLQSATGTT AYTQQMGMSV DMSSYQNTTS NLPQLAGFPV TVPAHPVAQQ HTNYHQQPLL.

The protein encoded by this gene is closely related to STAM, an adaptor protein involved in the downstream signaling of cytokine receptors, both of which contain a SH3 domain and the immunoreceptor tyrosine-based activation motif (ITAM). Similar to STAM, this protein acts downstream of JAK kinases, and is phosphorylated in response to cytokine stimulation. This protein and STAM thus are thought to exhibit compensatory effects on the signaling pathway downstream of JAK kinases upon cytokine stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAM2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STAM2
Gene Accession No. O75886, O88811, Q5XHY7
Gene Alias 1200004O12Rik; 5730456G07Rik; BB145015; DKFZp564C047; Hbp; HGNC:11358; Hrs binding protein; hrs-binding protein; HSE1 homolog; signal transducing adapter molecule 2; signal transducing adaptor molecule (SH3 domain and ITAM motif) 2; signal transducing adaptor molecule 2; STAM2; STAM-2; STAM2A; STAM2B; STAM-like protein containing SH3 and ITAM domains 2
Gene Symbols STAM2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 376-525 of human STAM2 (NP_005834.4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10254, 311030, 56324
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.