Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583276
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583276 Supplier Invitrogen™ Supplier No. PA583276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAPVYSVYSK LHPPAHYPPA SSGVPMQTYP VQSHGGNYMG QSIHQVTVAQ SYSLGPDQIG PLRSLPPNVN SSVTA Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 80%.

The protein encoded by this gene is closely related to STAM, an adaptor protein involved in the downstream signaling of cytokine receptors, both of which contain a SH3 domain and the immunoreceptor tyrosine-based activation motif (ITAM). Similar to STAM, this protein acts downstream of JAK kinases, and is phosphorylated in response to cytokine stimulation. This protein and STAM thus are thought to exhibit compensatory effects on the signaling pathway downstream of JAK kinases upon cytokine stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STAM2
Gene Accession No. O75886
Gene Alias 1200004O12Rik; 5730456G07Rik; BB145015; DKFZp564C047; Hbp; HGNC:11358; Hrs binding protein; hrs-binding protein; HSE1 homolog; signal transducing adapter molecule 2; signal transducing adaptor molecule (SH3 domain and ITAM motif) 2; signal transducing adaptor molecule 2; STAM2; STAM-2; STAM2A; STAM2B; STAM-like protein containing SH3 and ITAM domains 2
Gene Symbols STAM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STAM2. Recombinant protein control fragment (Product #RP-96005).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10254
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.