Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAM2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501568761
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8761 Supplier Bethyl Laboratories, Inc Supplier No. A304818A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

STAM contains an SH3 (Src homology 3) motif as well as an immunoreceptor tyrosine-based activation (ITAM) motif, both of which appear to be required for c-Myc induction in response to IL-2 and GM-CSF. STAM associates with JAK3 and JAK2 via its ITAM region, and it is tyrosine phosphorylated by JAK3 and JAK2 after stimulation with IL-2 and GM-CSF, respectively. STAM2, also known as Hbp, is a protein that is highly related to STAM. Similar to STAM, STAM2 functions downstream of JAK kinases and can be phosphorylated in response to cytokines. Due to alternative splicing events, two isoforms of STAM2 exist, namely STAM2A and STAM2B.

Specifications

Antigen STAM2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene STAM2
Gene Accession No. O75886
Gene Alias STAM2; signal transducing adaptor molecule (SH3 domain and ITAM motif) 2 ; HGNC:11358; DKFZp564C047; Hbp; STAM2A; STAM2B ; STAM-like protein containing SH3 and ITAM domains 2; signal transducing adaptor molecule 2
Gene Symbols STAM2
Host Species Rabbit
Immunogen Between 175 and 225
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10254
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.