Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STAMBPL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559103
Description
Immunogen sequence: LKKYNVEYQE YLQSKNKYKA EILKKLEHQR LIEAERKRIA QMRQQQLESE QFLFFEDQLK KQELARGQMR SQQTSGLS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
STAMBPL1 is zinc metalloprotease that specifically cleaves 'Lys-63'-linked polyubiquitin chains. Does not cleave 'Lys-48'-linked polyubiquitin chains.
Specifications
STAMBPL1 | |
Polyclonal | |
Unconjugated | |
STAMBPL1 | |
1700095N21Rik; 8230401J17Rik; ALMalpha; AMSH family protein; AMSH-family protein; AMSH-FP; AMSH-like protease; Amshlp; AMSH-LP; associated molecule with the SH3 domain of STAM (AMSH) - Family Protein; associated molecule with the SH3 domain of STAM (AMSH) like protein; bA399O19.2; KIAA1373; STAM binding protein like 1; STAM binding protein-like 1; STAM-binding protein-like 1; STAMBPL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57559 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96FJ0 | |
STAMBPL1 | |
Recombinant protein corresponding to Human STAMBPL1. Recombinant protein control fragment (Product #RP-95840). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction