Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STAMBPL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110438
Description
Immunogen sequence: ELKNDLLKKY NVEYQEYLQS KNKYKAEILK KLEHQRLIEA ERKRIAQMRQ QQLESEQFLF FEDQLKKQEL ARGQMRSQQT SGLSEQIDGS ALSCFSTHQN NSLLNVFADQ PNKSDATNYA SHSPPVNRAL T.
STAMBPL1 is zinc metalloprotease that specifically cleaves 'Lys-63'-linked polyubiquitin chains. Does not cleave 'Lys-48'-linked polyubiquitin chains.
Specifications
STAMBPL1 | |
Polyclonal | |
Unconjugated | |
STAMBPL1 | |
1700095N21Rik; 8230401J17Rik; ALMalpha; AMSH family protein; AMSH-family protein; AMSH-FP; AMSH-like protease; Amshlp; AMSH-LP; associated molecule with the SH3 domain of STAM (AMSH) - Family Protein; associated molecule with the SH3 domain of STAM (AMSH) like protein; bA399O19.2; KIAA1373; STAM binding protein like 1; STAM binding protein-like 1; STAM-binding protein-like 1; STAMBPL1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
687696, 76630 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.19 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q76N33 | |
STAMBPL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 120-250 of human STAMBPL1 (NP_065850.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction