Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAMBPL1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735639 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5639 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5639 Supplier Proteintech Group Inc Supplier No. 273151AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STAMBPL1 is zinc metalloprotease that specifically cleaves ′Lys-63′-linked polyubiquitin chains. Does not cleave ′Lys-48′-linked polyubiquitin chains.

Specifications

Antigen STAMBPL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene STAMBPL1
Gene Accession No. Q96FJ0
Gene Alias ALMalpha, AMSH FP, AMSH like protease, AMSH LP, AMSHLP, KIAA1373, STAM binding protein like 1, STAMBPL1
Gene Symbols STAMBPL1
Host Species Rabbit
Immunogen STAMBPL1 Fusion Protein Ag26308
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57559
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.