Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAMP Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567375
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7375 Supplier Bethyl Laboratories, Inc Supplier No. A304302A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan

Polyglutamylase which preferentially modifies alpha-tubulin. Involved in the side-chain initiation step of the polyglutamylation reaction rather than in the elongation step. Required for CCSAP localization to both spindle and cilia microtubules. Increases the effects of NCOA2 in glucocorticoid receptor-mediated repression and induction and in androgen receptor-mediated induction.

Specifications

Antigen STAMP
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TTLL5
Gene Accession No. Q6EMB2
Gene Alias KIAA0998, STAMP, SRC1 and TIF2 associated binding protein, SRC1 and TIF2-associated modulatory protein, tubulin polyglutamylase TTLL5
Gene Symbols TTLL5
Host Species Rabbit
Immunogen Between 825 and 875
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23093
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.