Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAMP2/STEAP4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | STAMP2/STEAP4 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
STAMP2/STEAP4 Polyclonal antibody specifically detects STAMP2/STEAP4 in Human samples. It is validated for ImmunofluorescenceSpecifications
STAMP2/STEAP4 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, pH 7.2, 40% glycerol | |
79689 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EC 1.16.1, EC 1.16.1.-, FLJ23153, Six-transmembrane epithelial antigen of prostate 4, SixTransMembrane protein of prostate 2, STAMP2DKFZp666D049, STEAP family member 4, TIARPsix transmembrane prostate protein 2, TNFAIP9, Tumor necrosis factor, alpha-induced protein 9metalloreductase STEAP4, tumor necrosis-alpha-induced adipose-related protein | |
This antibody was developed against Recombinant Protein corresponding to amino acids: TCIDALPLTMNSSEKQETVCIFGTGDFGRSLGLKMLQCGYSVVFGSRNPQKTTLLPSGAEVLSYSEAAKKSDIIIIAIHREHY | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title